Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Fluoro-3-methylphenyl)propionic acid, 97%

$209.45 - $209.45

Chemical Identifiers

Chemical Name or Material 3-(4-Fluoro-3-methylphenyl)propionic acid
CAS 881189-64-6
Molecular Formula C10H11FO2
Formula Weight 182.19
MDL Number MFCD09025393
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3247403

  SDS
alfa aesar™
H3247403
1g Each for $209.45
Description & Specifications

Specifications

Formula Weight 182.19
Assay 97%
Molecular Formula C10H11FO2
CAS 881189-64-6
Quantity 1g
Chemical Name or Material 3-(4-Fluoro-3-methylphenyl)propionic acid
Melting Point 64° to 69°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD09025393
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.