Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Fluoro-3,5-dimethylphenyl)propionic acid, 97%

$79.73 - $266.30

Chemical Identifiers

Chemical Name or Material 3-(4-Fluoro-3,5-dimethylphenyl)propionic acid
CAS 1017779-63-3
Molecular Formula C11H13FO2
Formula Weight 196.22
MDL Number MFCD09832377
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3157603

 
alfa aesar™
H3157603
1g Each for $266.30

AAH31576MD

 
alfa aesar™
H31576MD
250mg Each for $79.73
Description & Specifications

Specifications

Formula Weight 196.22
Chemical Name or Material 3-(4-Fluoro-3,5-dimethylphenyl)propionic acid
Molecular Formula C11H13FO2
MDL Number MFCD09832377
CAS 1017779-63-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Assay 97%
Melting Point 77.5° to 79°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.