missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Ethylenedioxythiophene, TCI America™

Manufacturer:  TCI America E07415G

 View more versions of this product

Catalog No. E07415G



Specifications

Specifications

3,4-Ethylenedioxythiophene
Undesignated
126213-50-1
MFCD02093622
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane, 2,3-dihydrothieno 3,4-b-1,4-dioxin
C1COC2=CSC=C2O1
142.172
142.17
≥98.0% (GC)
112°C
5g
C6H6O2S
2810
GKWLILHTTGWKLQ-UHFFFAOYSA-N
2,3-dihydrothieno[3,4-b][1,4]dioxine
4421864
Clear Liquid at 20°C

Chemical Identifiers

126213-50-1
142.172
GKWLILHTTGWKLQ-UHFFFAOYSA-N
4421864
C1COC2=CSC=C2O1
C6H6O2S
MFCD02093622
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2h,3h-thiopheno 3,4-e 1,4-dioxane, 2,3-dihydrothieno 3,4-b-1,4-dioxin
2,3-dihydrothieno[3,4-b][1,4]dioxine
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6421

TSCA : Yes

Recommended Storage : Refrigerator

Documents