Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Ethoxyphenyl)propionic acid, 98%

$38.18 - $143.78

Chemical Identifiers

Chemical Name or Material 3-(4-Ethoxyphenyl)propionic acid
CAS 4919-34-0
Molecular Formula C11H14O3
Formula Weight 194.23
MDL Number MFCD00016557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5021403

 
alfa aesar™
H5021403
1g Each for $38.18

AAH5021406

 
alfa aesar™
H5021406
5g Each for $143.78
Description & Specifications

Specifications

Formula Weight 194.23
Chemical Name or Material 3-(4-Ethoxyphenyl)propionic acid
Assay 98%
Molecular Formula C11H14O3
MDL Number MFCD00016557
CAS 4919-34-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.