Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dinitrobenzonitrile, 97%

$64.34 - $64.34

Chemical Identifiers

Chemical Name or Material 3,4-Dinitrobenzonitrile
CAS 4248-33-3
Molecular Formula C7H3N3O4
Formula Weight 193.12
MDL Number MFCD00013370
UN Number UN3439
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1313103

  SDS
alfa aesar™
A1313103
1g Each for $64.34

AAA1313106

  SDS
alfa aesar™
A1313106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 193.12
CAS 4248-33-3
Quantity 1g
MDL Number MFCD00013370
Molecular Formula C7H3N3O4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 88° to 92°C
UN Number UN3439
Assay 97%
Chemical Name or Material 3,4-Dinitrobenzonitrile
Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H373-H312-H332-H315-H319
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.