Learn More
3,4-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1877406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,4-Dinitrobenzoic acid | |
163°C to 167°C | |
MFCD00007255 | |
2055333 | |
3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956 | |
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O | |
212.12 | |
212.12 |
528-45-0 | |
C7H4N2O6 | |
5 g | |
14,3275 | |
OMVRRHJJQILIJX-UHFFFAOYSA-N | |
3,4-dinitrobenzoic acid | |
10709 | |
99% |
Chemical Identifiers
528-45-0 | |
212.12 | |
OMVRRHJJQILIJX-UHFFFAOYSA-N | |
10709 | |
OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O |
C7H4N2O6 | |
MFCD00007255 | |
3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956 | |
3,4-dinitrobenzoic acid |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 208-433-9
RTECSNumber : DG9140650
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only