Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylthiophene-2,5-dicarbonitrile, 98%

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylthiophene-2,5-dicarbonitrile
CAS 155632-41-0
Molecular Formula C8H6N2S
Formula Weight 162.21
MDL Number MFCD00041438
UN Number UN3439
Beilstein 6799021
Synonym 2,5-Dicyano-3,4-dimethylthiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2437103

  SDS
alfa aesar™
B2437103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2437106

  SDS
alfa aesar™
B2437106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.21
Chemical Name or Material 3,4-Dimethylthiophene-2,5-dicarbonitrile
Synonym 2,5-Dicyano-3,4-dimethylthiophene
CAS 155632-41-0
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3439
Beilstein 6799021
Assay 98%
Melting Point 122° to 124°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Molecular Formula C8H6N2S
Quantity 1g
MDL Number MFCD00041438
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.