Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylphenylhydrazine hydrochloride, 98%

$79.72 - $1,365.12

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylphenylhydrazine hydrochloride
CAS 60481-51-8
Molecular Formula C8H12N2·HCl
Formula Weight 172.66
MDL Number MFCD00052270
Beilstein 6096060
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1302709

  SDS
alfa aesar™
A1302709
10g Each for $79.72

AAA1302730

  SDS
alfa aesar™
A1302730
250g Each for $1,365.12
Description & Specifications

Specifications

Formula Weight 172.66
Chemical Name or Material 3,4-Dimethylphenylhydrazine hydrochloride
Assay 98%
Molecular Formula C8H12N2·HCl
Melting Point ≈195°C (dec.)
MDL Number MFCD00052270
CAS 60481-51-8
Beilstein 6096060
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,4-Dimethylphenylhydrazine hydrochloride, is used as a reactant with ethyl 3-ethoxy-2-ethoxycarbonylprop-2-enoate to produce 2-(3,4-dimethyl-phenyl)-3-oxo-2,3-dihydro-1H-pyrazole-4-carboxylic acid ethyl ester. This reaction will need reagent K2CO3, and the menstruum ethanol.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.