missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethylphenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 55499-43-9
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD01009694
InChI Key KDVZJKOYSOFXRV-UHFFFAOYSA-N
Synonym 3,4-dimethyl phenylboronic acid
PubChem CID 2734348
IUPAC Name (3,4-dimethylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)C)(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359280050 SDS View Documents Thermo Fisher Scientific
359280050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

55499-43-9
149.984
KDVZJKOYSOFXRV-UHFFFAOYSA-N
2734348
B(C1=CC(=C(C=C1)C)C)(O)O
C8H11BO2
MFCD01009694
3,4-dimethyl phenylboronic acid
(3,4-dimethylphenyl)boronic acid

Specifications

Authentic
White to Beige
55499-43-9
100.0
C8H11BO2
3,4-dimethyl phenylboronic acid
B(C1=CC(=C(C=C1)C)C)(O)O
149.984
149.98
97%
Glass bottle
5g
96.0
97%
MFCD01009694
KDVZJKOYSOFXRV-UHFFFAOYSA-N
(3,4-dimethylphenyl)boronic acid
2734348
Powder
3, 4-Dimethylphenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.