Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylphenoxyacetic acid, 98%

$64.34 - $253.14

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylphenoxyacetic acid
CAS 13335-73-4
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00017595
Beilstein 2558866
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1535506

  SDS
alfa aesar™
A1535506
5g Each for $64.34

AAA1535514

  SDS
alfa aesar™
A1535514
25g Each for $253.14
Description & Specifications

Specifications

Formula Weight 180.2
Assay 98%
CAS 13335-73-4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00017595
Melting Point 160° to 165°C
Beilstein 2558866
Quantity 5g
Molecular Formula C10H12O3
Chemical Name or Material 3,4-Dimethylphenoxyacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.