missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylphenoxyacetic acid, 98%

Manufacturer:  Alfa Aesar™ A1535506

 View more versions of this product

Catalog No. AAA1535506



Specifications

Specifications

3,4-Dimethylphenoxyacetic acid
5g
C10H12O3
2558866
GIEQJKZWTIFEJM-UHFFFAOYSA-N
2-(3,4-dimethylphenoxy)acetic acid
83355
98%
160°C to 165°C
13335-73-4
MFCD00017595
2-3,4-dimethylphenoxy acetic acid, 3,4-dimethylphenoxyacetic acid, 3,4-dimethylphenoxy acetic acid, 3,4-xylyloxyacetic acid, acmc-1bqwp, 3,4-dimethyl-phenoxy acetic acid, 3,4-dimethyl-phenoxy-acetic acid, acetic acid,2-3,4-dimethylphenoxy, acetic acid, 2-3,4-dimethylphenoxy
CC1=C(C=C(C=C1)OCC(=O)O)C
180.203
180.2

Chemical Identifiers

13335-73-4
180.203
GIEQJKZWTIFEJM-UHFFFAOYSA-N
83355
CC1=C(C=C(C=C1)OCC(=O)O)C
C10H12O3
MFCD00017595
2-3,4-dimethylphenoxy acetic acid, 3,4-dimethylphenoxyacetic acid, 3,4-dimethylphenoxy acetic acid, 3,4-xylyloxyacetic acid, acmc-1bqwp, 3,4-dimethyl-phenoxy acetic acid, 3,4-dimethyl-phenoxy-acetic acid, acetic acid,2-3,4-dimethylphenoxy, acetic acid, 2-3,4-dimethylphenoxy
2-(3,4-dimethylphenoxy)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents