Learn More
3,4-Dimethylphenol, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116580025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 4-Dimethylphenol | |
95-65-8 | |
100.0 | |
227°C | |
Authentic | |
Glass bottle | |
(CH3)2C6H3OH | |
2.5kg | |
0.983 | |
3,4-xylenol, phenol, 3,4-dimethyl, 4,5-dimethylphenol, 3,4-dimethyl phenol, 1,3,4-xylenol, 4-hydroxy-1,2-dimethylbenzene, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl-phenol, 3,4-dmp, ccris 723 | |
YCOXTKKNXUZSKD-UHFFFAOYSA-N | |
3,4-dimethylphenol | |
7249 | |
122.17 |
99% | |
98.5 | |
0.9830g/mL | |
80°C | |
98.5% min. (GC) | |
C8H10O | |
MFCD00002304 | |
06, 480 | |
15, 10279 | |
Solubility in water: slightly soluble. Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene | |
CC1=CC=C(O)C=C1C | |
122.17 | |
CHEBI:39839 | |
99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 202-439-5
RTECSNumber : ZE6300000
TSCA : TSCA
RUO â Research Use Only