Learn More
3,4-Dimethylphenol, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 116585000
Specifications
3, 4-Dimethylphenol | |
Glass bottle | |
63°C to 67°C | |
99% | |
98.5 | |
99% | |
(CH3)2C6H3OH | |
06, 480 | |
1,3,4-xylenol, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl phenol, 3,4-dimethyl-phenol, 3,4-dmp, 3,4-xylenol, 4,5-dimethylphenol, 4-hydroxy-1,2-dimethylbenzene, ccris 723, phenol, 3,4-dimethyl | |
CC1=C(C=C(C=C1)O)C | |
122.167 | |
CHEBI:39839 | |
Crystalline Powder |
Authentic | |
Cream to White | |
500g | |
95-65-8 | |
100.0 | |
C8H10O | |
MFCD00002304 | |
15, 10279 | |
YCOXTKKNXUZSKD-UHFFFAOYSA-N | |
3,4-dimethylphenol | |
7249 | |
122.17 | |
≥98.5% (GC) |
Chemical Identifiers
95-65-8 | |
122.167 | |
YCOXTKKNXUZSKD-UHFFFAOYSA-N | |
7249 | |
3,4-dimethylphenol |
C8H10O | |
MFCD00002304 | |
1,3,4-xylenol, 1-hydroxy-3,4-dimethylbenzene, 3,4-dimethyl phenol, 3,4-dimethyl-phenol, 3,4-dmp, 3,4-xylenol, 4,5-dimethylphenol, 4-hydroxy-1,2-dimethylbenzene, ccris 723, phenol, 3,4-dimethyl | |
CHEBI:39839 | |
CC1=C(C=C(C=C1)O)C |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-439-5
RTECSNumber : ZE6300000
TSCA : TSCA