Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylphenol, 98+%

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylphenol
CAS 95-65-8
Molecular Formula C8H10O
Formula Weight 122.17
MDL Number MFCD00002304
UN Number UN2261
Beilstein 1099267
Merck Index 14,10082
Synonym 3,4-Xylenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1059922

  SDS
alfa aesar™
A1059922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Merck Index 14,10082
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
MDL Number MFCD00002304
TSCA TSCA
Beilstein 1099267
UN Number UN2261
Synonym 3,4-Xylenol
RTECS Number ZE6300000
Melting Point 65° to 68°C
Health Hazard 2 GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Quantity 100g
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3,4-Dimethylphenol
Assay ≥98%
Molecular Formula C8H10O
CAS 95-65-8
Boiling Point 227°C
Health Hazard 1 Danger

Applications
3,4-Dimethylphenol, is used as an anti-Infective agents. It has wide applications in the analytical techniques such as Gas Chromatography, Reversed Phase Chromatography, HPLC and UHPLC.

Solubility
Slightly soluble in water. Less than 1 mg/mL at 68° F.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive.Hygroscopic. Incompatible with bases, acid chlorides, acid anhydrides, and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H310-H314-H318-H302
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.