missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylhexane, 97%

Manufacturer:  Alfa Aesar™ H5349806

 View more versions of this product

Catalog No. AAH5349806



Description

Description

Applications
3,4-Dimethylhexane may be used to study the mechanism of addition of hydrogen to 1-butene.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

3,4-Dimethylhexane
6°C (43°F)
119°C
583-48-2
MFCD00039927
1718744
RNTWWGNZUXGTAX-UHFFFAOYSA-N
3,4-dimethylhexane
11412
97%
0.72
1.403
5g
C8H18
UN1262
3,4-dimethyl hexane
CCC(C)C(C)CC
114.232
114.23

Chemical Identifiers

583-48-2
114.232
RNTWWGNZUXGTAX-UHFFFAOYSA-N
11412
CCC(C)C(C)CC
C8H18
MFCD00039927
3,4-dimethyl hexane
3,4-dimethylhexane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES

EINECSNumber : 209-504-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents