Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylbenzyl alcohol, 97%

$52.49 - $729.62

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylbenzyl alcohol
CAS 6966-10-5
Molecular Formula C9H12O
Formula Weight 136.19
MDL Number MFCD00004647
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2492603

  SDS
alfa aesar™
B2492603
1g Each for $52.49

AAB2492606

  SDS
alfa aesar™
B2492606
5g Each for $175.47

AAB2492614

  SDS
alfa aesar™
B2492614
25g Each for $729.62
Description & Specifications

Specifications

Formula Weight 136.19
Chemical Name or Material 3,4-Dimethylbenzyl alcohol
Assay 97%
Molecular Formula C9H12O
Melting Point 62° to 65°C
Boiling Point 218° to 221°C
MDL Number MFCD00004647
CAS 6966-10-5
Quantity 1g