Please login to your online account to display your discounted pricing

3,4-Dimethylbenzoic Acid 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Dimethylbenzoic acid
Name Note 98%
CAS 619-04-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H10O2
Linear Formula (CH3)2C6H3CO2H
Formula Weight 150.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115990100

  SDS
thermo fisher scientific
AC115990100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC115991000

  SDS
thermo fisher scientific
AC115991000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 150.18
Quantity 10g
Color White to beige
TSCA TSCA
RTECS Number DG8734020
EINECS Number 210-576-7
Melting Point 162° to 168°C
CAS Max % 100.0
Health Hazard 1 Warning
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
CAS Min % 97.5
Synonym asym-o-Xylylic acid
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Molecular Formula C9H10O2
Linear Formula (CH3)2C6H3CO2H
CAS 619-04-5
Chemical Name or Material 3, 4-Dimethylbenzoic acid
Assay Percent Range 98%
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.