missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylbenzamide, 98%

Manufacturer:  Alfa Aesar™ A1199503

 View more versions of this product

Catalog No. AAA1199503


 

Specifications

Specifications

3,4-Dimethylbenzamide
1g
98%
MFCD00052960
CC1=C(C=C(C=C1)C(=O)N)C
149.193
149.19
101°C to 103°C
5580-33-6
C9H11NO
INGCXEIJXKQPJH-UHFFFAOYSA-N
3,4-dimethylbenzamide
21755

Chemical Identifiers

5580-33-6
149.193
INGCXEIJXKQPJH-UHFFFAOYSA-N
C9H11NO
MFCD00052960
21755
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents