Please login to your online account to display your discounted pricing

3,4-Dimethylbenzaldehyde, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  346060050

 View more versions of this product

Catalog No. AC346060050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 134.18
Quantity 5g
Color Clear colorless to yellow
EINECS Number 227-770-2
TSCA TSCA
Density 1.0120g/mL
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 97%
Molecular Formula C9H10O
MDL Number MFCD00016612
CAS 5973-71-7
Health Hazard 1 AIR SENSITIVE
Packaging Glass bottle
Refractive Index 1.5490 to 1.5510 (20°C, 589nm)
Chemical Name or Material 3, 4-Dimethylbenzaldehyde
Linear Formula (CH3)2C6H3CHO

Specifications

Chemical Name or Material 3, 4-Dimethylbenzaldehyde
Name Note 98%
CAS 5973-71-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O
Linear Formula (CH3)2C6H3CHO
Formula Weight 134.18
MDL Number MFCD00016612
Accessories

AIR SENSITIVE

SDS & Safety Documents