missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethylbenzaldehyde, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 346060250

 View more versions of this product

Catalog No. AC346060250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

134.18
≥96.0%
Clear colorless to yellow
100.0
Authentic
Glass bottle
5973-71-7
C9H10O
97%
Liquid
25g
1.0120g/mL
96.0
1.5490 to 1.5510 (20°C, 589nm)
(CH3)2C6H3CHO
3, 4-Dimethylbenzaldehyde
MFCD00016612
98%

Chemical Identifiers

5973-71-7
100.0
C9H10O
134.18
3, 4-Dimethylbenzaldehyde, 98%
96.0
97%
(CH3)2C6H3CHO
MFCD00016612
≥96.0%

Safety and Handling

SDS & Safety Documents

AIR SENSITIVE

EINECSNumber : 227-770-2

TSCA : TSCA