missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1538806

 View more versions of this product

Catalog No. AAA1538806



Description

Description

3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzaldehyde is used as an additive for resins, agricultural intermediate and for flavor and fragrance, nucleation transparent agent.

Solubility
Soluble in ethanol, ether, toluene. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Keep way from oxidizing agents, air.
Specifications

Specifications

3,4-Dimethylbenzaldehyde
1.017
Almond-like
1.551
5 g
Air Sensitive
Soluble in ethanol,ether,toluene. Insoluble in water.
CC1=C(C=C(C=C1)C=O)C
134.178
134.18
5973-71-7
226°C
C9H10O
MFCD00016612
1634270
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
POQJHLBMLVTHAU-UHFFFAOYSA-N
3,4-dimethylbenzaldehyde
22278
97%

Chemical Identifiers

5973-71-7
134.178
POQJHLBMLVTHAU-UHFFFAOYSA-N
22278
CC1=C(C=C(C=C1)C=O)C
C9H10O
MFCD00016612
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
3,4-dimethylbenzaldehyde
Safety and Handling

Safety and Handling

EINECSNumber : 227-770-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only