missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylaniline 99.0+%, TCI America™

Supplier:  TCI America D067025G

 View more versions of this product

Catalog No. D067025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,4-Dimethylaniline
95-64-7
25 g
C8H11N
3452
DOLQYFPDPKPQSS-UHFFFAOYSA-N
3,4-dimethylaniline
7248
121.18
51°C
226°C
Crystal-Powder at 20°C
MFCD00007810
3,4-xylidine, 4-amino-o-xylene, 3,4-xylylamine, 3,4-dimethylphenylamine, benzenamine, 3,4-dimethyl, 3,4-dimethylbenzenamine, 3,4-dimethylaminobenzene, 1-amino-3,4-dimethylbenzene, 4-amino-1,2-dimethylbenzene, aniline, 3,4-dimethyl
CC1=CC=C(N)C=C1C
121.18
CHEBI:39901
≥99.0% (GC,T)

Chemical Identifiers

95-64-7
121.18
DOLQYFPDPKPQSS-UHFFFAOYSA-N
7248
3,4-dimethylaniline
C8H11N
MFCD00007810
3,4-xylidine, 4-amino-o-xylene, 3,4-xylylamine, 3,4-dimethylphenylamine, benzenamine, 3,4-dimethyl, 3,4-dimethylbenzenamine, 3,4-dimethylaminobenzene, 1-amino-3,4-dimethylbenzene, 4-amino-1,2-dimethylbenzene, aniline, 3,4-dimethyl
CHEBI:39901
CC1=CC=C(N)C=C1C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0129

RTECSNumber : ZE9450000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions