missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethylaniline, 99.5%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 155285000

 View more versions of this product

Catalog No. AC155285000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 4-Dimethylaniline
Authentic
226°C
49°C to 51°C
99.5%
99.4
99.4% min. (GC)
(CH3)2C6H3NH2
12, 1103
1-amino-3,4-dimethylbenzene
DOLQYFPDPKPQSS-UHFFFAOYSA-N
3,4-dimethylaniline
7248
121.18
99.50%
98°C
Glass bottle
Beige to Brown
500g
95-64-7
100.0
C8H11N
MFCD00007810
15, 10282
Solubility in water: <1g/L (24°C). Other solubilities: slightly soluble in chloroform,soluble in alcohol (>100g/L),diethyl ether,,petroleum ether,very soluble in ligroin
CC1=C(C=C(C=C1)N)C
121.18
CHEBI:39901
Crystalline Mass or Crystals

Chemical Identifiers

95-64-7
121.18
DOLQYFPDPKPQSS-UHFFFAOYSA-N
7248
3,4-dimethylaniline
C8H11N
MFCD00007810
1-amino-3,4-dimethylbenzene
CHEBI:39901
CC1=C(C=C(C=C1)N)C
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 202-437-4

RTECSNumber : ZE9450000

TSCA : TSCA

Recommended Storage : product darkens if exposed to light and air

SDS Documents