Please login to your online account to display your discounted pricing

3,4-Dimethylaniline, 99.5%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4-Dimethylaniline
Name Note 99.5%
CAS 95-64-7
CAS Min % 99.4
CAS Max % 100.0
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C8H11N
Linear Formula (CH3)2C6H3NH2
Formula Weight 121.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC155280050

  SDS
thermo fisher scientific
155280050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155281000

  SDS
thermo fisher scientific
155281000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC155285000

  SDS
thermo fisher scientific
155285000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Mass or Crystals
Assay ≥99.4%
Formula Weight 121.18
Quantity 5g
Color Light beige to pink to brown
EINECS Number 202-437-4
TSCA TSCA
RTECS Number ZE9450000
Melting Point 49° to 51°C
Beilstein 12, 1103
Merck Index 15, 10282
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Storage Information product darkens if exposed to light and air
Synonym 3, 4-Xylidine
CAS Min % 99.4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 4-Dimethylaniline
Name Note 99.5%
Assay Percent Range 99.5%
Molecular Formula C8H11N
Linear Formula (CH3)2C6H3NH2
MDL Number MFCD00007810
CAS 95-64-7
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic if swallowed.