Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethylaniline, 98%

Chemical Identifiers

Chemical Name or Material 3,4-Dimethylaniline
CAS 95-64-7
Molecular Formula C8H11N
Formula Weight 121.18
MDL Number MFCD00007810
UN Number UN3452
Beilstein 507414
Merck Index 14,10084
Synonym 3,4-Xylidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA169280B

  SDS
alfa aesar™
A169280B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1692822

  SDS
alfa aesar™
A1692822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1692830

  SDS
alfa aesar™
A1692830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.18
Chemical Name or Material 3,4-Dimethylaniline
TSCA TSCA
Assay 98%
Molecular Formula C8H11N
MDL Number MFCD00007810
CAS 95-64-7
UN Number UN3452
Beilstein 507414
Merck Index 14,10084
Synonym 3,4-Xylidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, SOLID
RTECS Number ZE9450000
Flash Point 98°C (208°F)
Odor Characteristic
Boiling Point 226° to 228°C
Melting Point 48° to 52°C
Quantity 1000g

Applications
3,4-Dimethylaniline is used to study electron donor-acceptor interaction with DCNQ. It is also used as a fine chemical &organic intermediate.

Solubility
Soluble in water and in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates, halogens.

Safety and Handling

Danger

GHS P Statement
P280h-P273-P309-P310-P302+P352
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.