missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4'-Dimethylacetanilide, 98+%

$53.66 - $172.72

Chemical Identifiers

CAS 2198-54-1
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00026137
InChI Key UAOIEEWQVAXCFY-UHFFFAOYSA-N
Synonym 3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
PubChem CID 75144
IUPAC Name N-(3,4-dimethylphenyl)acetamide
SMILES CC1=C(C=C(C=C1)NC(=O)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0191906 View Documents Alfa Aesar™
L0191906
5g Each for $53.66
 
AAL0191914 View Documents Alfa Aesar™
L0191914
25g Each for $172.72
 
Specifications

Chemical Identifiers

2198-54-1
163.22
UAOIEEWQVAXCFY-UHFFFAOYSA-N
75144
CC1=C(C=C(C=C1)NC(=O)C)C
C10H13NO
MFCD00026137
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
N-(3,4-dimethylphenyl)acetamide

Specifications

96°C to 98°C
2198-54-1
MFCD00026137
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
CC1=C(C=C(C=C1)NC(=O)C)C
163.22
163.22
3',4'-Dimethylacetanilide
5g
C10H13NO
387706
UAOIEEWQVAXCFY-UHFFFAOYSA-N
N-(3,4-dimethylphenyl)acetamide
75144
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AN7500000