missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3',4'-Dimethylacetanilide, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0191914
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3',4'-Dimethylacetanilide | |
96°C to 98°C | |
MFCD00026137 | |
387706 | |
UAOIEEWQVAXCFY-UHFFFAOYSA-N | |
N-(3,4-dimethylphenyl)acetamide | |
75144 | |
≥98% |
2198-54-1 | |
C10H13NO | |
25 g | |
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide | |
CC1=C(C=C(C=C1)NC(=O)C)C | |
163.22 | |
163.22 |
Chemical Identifiers
2198-54-1 | |
163.22 | |
UAOIEEWQVAXCFY-UHFFFAOYSA-N | |
75144 | |
CC1=C(C=C(C=C1)NC(=O)C)C |
C10H13NO | |
MFCD00026137 | |
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide | |
N-(3,4-dimethylphenyl)acetamide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 218-597-3
RTECSNumber : AN7500000
TSCA : No
Recommended Storage : Ambient temperatures