missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3709906
Description
3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
3,4-Dimethyl-1H-pyrazole | |
53°C to 55°C | |
C5H8N2 | |
5 g | |
Slightly soluble in water. | |
CC1=C(NN=C1)C | |
96.133 | |
96.13 | |
Fused Solid |
2820-37-3 | |
Cream | |
MFCD00462154 | |
3,4-dimethyl-1h-pyrazole, 3 5 ,4-dimethylpyrazole, 3,4-dimethylpyrazole, 3,4-dimethyl pyrazole, 1h-pyrazole, 3,4-dimethyl, unii-7lf1k5quj0, 7lf1k5quj0, 1h-pyrazole,4,5-dimethyl, 1h-pyrazole, 4,5-dimethyl, pyrazole, 3,4-dimethyl | |
VQTVFIMEENGCJA-UHFFFAOYSA-N | |
4,5-dimethyl-1H-pyrazole | |
137735 | |
97% |
Chemical Identifiers
2820-37-3 | |
96.133 | |
VQTVFIMEENGCJA-UHFFFAOYSA-N | |
137735 | |
CC1=C(NN=C1)C |
C5H8N2 | |
MFCD00462154 | |
3,4-dimethyl-1h-pyrazole, 3 5 ,4-dimethylpyrazole, 3,4-dimethylpyrazole, 3,4-dimethyl pyrazole, 1h-pyrazole, 3,4-dimethyl, unii-7lf1k5quj0, 7lf1k5quj0, 1h-pyrazole,4,5-dimethyl, 1h-pyrazole, 4,5-dimethyl, pyrazole, 3,4-dimethyl | |
4,5-dimethyl-1H-pyrazole |
Safety and Handling
GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
EINECSNumber : 429-130-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only