missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethyl-1,2-cyclopentanedione, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265050025

 View more versions of this product

Catalog No. AC265050025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dimethyl-1, 2-cyclopentanedione
Authentic
Yellow to Beige
2.5g
13494-06-9
100.0
C7H10O2
1,2-cyclopentanedione, 3,4-dimethyl, 1,2-cyclopentanedione,3,4-dimethyl, 3,4-dimethyl 1,2-cyclopentandione, 3,4-dimethyl-1,2-cyclopentadione, 3,4-dimethyl-1,2-cyclopentadione, fg, 3,4-dimethyl-1,2-cyclopentanedione, acmc-209bxe, caramel pentadione, fema no. 3268
WGAVDEVFJDQIMZ-UHFFFAOYSA-N
3,4-dimethylcyclopentane-1,2-dione
61633
Crystalline Powder
>100°C
Glass bottle
64°C to 69°C
97%
96.0
97%
MFCD00143063
Solubility in water: slightly soluble
CC1CC(=O)C(=O)C1C
126.15
126.15
97%

Chemical Identifiers

13494-06-9
126.15
WGAVDEVFJDQIMZ-UHFFFAOYSA-N
61633
CC1CC(=O)C(=O)C1C
C7H10O2
MFCD00143063
1,2-cyclopentanedione, 3,4-dimethyl, 1,2-cyclopentanedione,3,4-dimethyl, 3,4-dimethyl 1,2-cyclopentandione, 3,4-dimethyl-1,2-cyclopentadione, 3,4-dimethyl-1,2-cyclopentadione, fg, 3,4-dimethyl-1,2-cyclopentanedione, acmc-209bxe, caramel pentadione, fema no. 3268
3,4-dimethylcyclopentane-1,2-dione
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 236-810-8

TSCA : TSCA

SDS Documents