missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dimethyl-1,2-cyclopentanedione, 97%, ACROS Organics™
Chemical Identifiers
CAS | 13494-06-9 |
---|---|
Molecular Formula | C7H10O2 |
Molecular Weight (g/mol) | 126.15 |
MDL Number | MFCD00143063 |
InChI Key | WGAVDEVFJDQIMZ-UHFFFAOYSA-N |
Synonym | 1,2-cyclopentanedione, 3,4-dimethyl, 1,2-cyclopentanedione,3,4-dimethyl, 3,4-dimethyl 1,2-cyclopentandione, 3,4-dimethyl-1,2-cyclopentadione, 3,4-dimethyl-1,2-cyclopentadione, fg, 3,4-dimethyl-1,2-cyclopentanedione, acmc-209bxe, caramel pentadione, fema no. 3268 |
PubChem CID | 61633 |
IUPAC Name | 3,4-dimethylcyclopentane-1,2-dione |
SMILES | CC1CC(=O)C(=O)C1C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC265050025
|
Thermo Fisher Scientific
265050025 |
2.5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
13494-06-9 | |
126.15 | |
WGAVDEVFJDQIMZ-UHFFFAOYSA-N | |
61633 | |
CC1CC(=O)C(=O)C1C |
C7H10O2 | |
MFCD00143063 | |
1,2-cyclopentanedione, 3,4-dimethyl, 1,2-cyclopentanedione,3,4-dimethyl, 3,4-dimethyl 1,2-cyclopentandione, 3,4-dimethyl-1,2-cyclopentadione, 3,4-dimethyl-1,2-cyclopentadione, fg, 3,4-dimethyl-1,2-cyclopentanedione, acmc-209bxe, caramel pentadione, fema no. 3268 | |
3,4-dimethylcyclopentane-1,2-dione |
Specifications
>100°C | |
Glass bottle | |
64°C to 69°C | |
13494-06-9 | |
100.0 | |
C7H10O2 | |
1,2-cyclopentanedione, 3,4-dimethyl, 1,2-cyclopentanedione,3,4-dimethyl, 3,4-dimethyl 1,2-cyclopentandione, 3,4-dimethyl-1,2-cyclopentadione, 3,4-dimethyl-1,2-cyclopentadione, fg, 3,4-dimethyl-1,2-cyclopentanedione, acmc-209bxe, caramel pentadione, fema no. 3268 | |
WGAVDEVFJDQIMZ-UHFFFAOYSA-N | |
3,4-dimethylcyclopentane-1,2-dione | |
61633 | |
Crystalline Powder | |
3, 4-Dimethyl-1, 2-cyclopentanedione, 97% |
Authentic | |
Yellow to Beige | |
2.5g | |
96.0 | |
97% | |
MFCD00143063 | |
Solubility in water: slightly soluble | |
CC1CC(=O)C(=O)C1C | |
126.15 | |
126.15 | |
97% |
Safety and Handling
LIGHT SENSITIVE
EINECSNumber : 236-810-8
TSCA : TSCA