Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxythiophenol, 98%

$40.65 - $464.86

Chemical Identifiers

Chemical Name or Material 3,4-Dimethoxythiophenol
CAS 700-96-9
Molecular Formula C8H10O2S
Formula Weight 170.23
MDL Number MFCD00052823
Beilstein 1945136
Synonym 3,4-Dimethoxybenzenethiol; 4-Mercaptoveratrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1474503

  SDS
alfa aesar™
A1474503
1g Each for $40.65

AAA1474514

  SDS
alfa aesar™
A1474514
25g Each for $464.86

AAA1474506

  SDS
alfa aesar™
A1474506
5g Each for $153.78
Description & Specifications

Specifications

Formula Weight 170.23
Chemical Name or Material 3,4-Dimethoxythiophenol
Assay 98%
Molecular Formula C8H10O2S
MDL Number MFCD00052823
CAS 700-96-9
Beilstein 1945136
Synonym 3,4-Dimethoxybenzenethiol; 4-Mercaptoveratrole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point >113°C (235°F)
Odor Stench
Boiling Point 263°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.