Please login to your online account to display your discounted pricing

Alfa Aesar™ (3,4-Dimethoxyphenylthio)acetic acid, 97%

$75.78 - $75.78

Chemical Identifiers

Chemical Name or Material (3,4-Dimethoxyphenylthio)acetic acid
CAS 95735-63-0
Molecular Formula C10H12O4S
Formula Weight 228.26
MDL Number MFCD00052112
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2056903

  SDS
alfa aesar™
B2056903
1g Each for $75.78
Description & Specifications

Specifications

Formula Weight 228.26
Chemical Name or Material (3,4-Dimethoxyphenylthio)acetic acid
Assay 97%
Molecular Formula C10H12O4S
MDL Number MFCD00052112
CAS 95735-63-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 101° to 103°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.