Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxyphenylglyoxal hydrate, 98%, dry wt basis


Quantity 5g

Manufacturer: alfa aesar™  L2027906

 View more versions of this product

Catalog No. AAL2027906

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 194.19 (Anhydrous)
Chemical Name or Material 3,4-Dimethoxyphenylglyoxal hydrate
Health Hazard 1 Warning
Assay 98%
Molecular Formula C10H10O4·xH2O
MDL Number MFCD06800598
CAS 163428-90-8
Beilstein 3092619
Melting Point 118° to 120°C
Assay Percent Notes (dry wt.)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g

Specifications

Chemical Name or Material 3,4-Dimethoxyphenylglyoxal hydrate
CAS 163428-90-8
Molecular Formula C10H10O4·xH2O
Formula Weight 194.19 (Anhydrous)
MDL Number MFCD06800598
Beilstein 3092619
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.