3,4-Dimethoxyphenylacetone, 97%, Acros Organics™
Manufacturer:  Acros Organics AC161490050
 View more versions of this product
Catalog No. AC161490050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
3,4-Dimethoxyphenylacetone
>110°C
1.5338 to 1.5358
5g
C11H14O3
3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
CC(=O)CC1=CC(=C(C=C1)OC)OC
194.23
194.23
1.1150g/mL
Authentic
154.0°C to 158.0°C (12.0mmHg)
776-99-8
(CH3O)2C6H3CH2COCH3
UMYZWICEDUEWIM-UHFFFAOYSA-N
1-(3,4-dimethoxyphenyl)propan-2-one
69896
97%
Chemical Identifiers
776-99-8
194.23
3,4-dimethoxyphenylacetone, 1-3,4-dimethoxyphenyl propan-2-one, 3,4-dimethoxyphenyl acetone, veratryl-2-propanone, 1-3,4-dimethoxyphenyl-2-propanone, veratryl acetone, 3,4-dimethoxybenzyl methyl ketone, 1-3,4-dimethoxyphenyl acetone, 2-propanone, 1-3,4-dimethoxyphenyl, veratryl methyl ketone
1-(3,4-dimethoxyphenyl)propan-2-one
C11H14O3
UMYZWICEDUEWIM-UHFFFAOYSA-N
69896
CC(=O)CC1=CC(=C(C=C1)OC)OC
SDS Documents