missing translation for 'onlineSavingsMsg'
Learn More
missing translation for 'pleaseLogIn'

Alfa Aesar™ 3,4-Dimethoxyphenylacetic acid hydrazide, 97%

$30.32 - $330.95

Chemical Identifiers

CAS 60075-23-2
Molecular Formula C10H14N2O3
Formula Weight 210.23
MDL Number MFCD00051701
Beilstein 2698478
Synonym 3,4-Dimethoxyphenylacethydrazide; Homoveratric acid hydrazide
Chemical Name or Material 3,4-Dimethoxyphenylacetic acid hydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1104903 SDS Alfa Aesar™
A1104903
1g Each for $30.32
AAA1104906 SDS Alfa Aesar™
A1104906
5g Each for $83.58
AAA1104914 SDS Alfa Aesar™
A1104914
25g Each for $330.95
Specifications Description & Specifications

Chemical Identifiers

60075-23-2
210.23
2698478
3,4-Dimethoxyphenylacetic acid hydrazide
C10H14N2O3
MFCD00051701
3,4-Dimethoxyphenylacethydrazide; Homoveratric acid hydrazide

Specifications

102° to 104°C
60075-23-2
210.23
2698478
3,4-Dimethoxyphenylacetic acid hydrazide
1g
C10H14N2O3
MFCD00051701
3,4-Dimethoxyphenylacethydrazide; Homoveratric acid hydrazide
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning