missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxyphenyl Isothiocyanate, TCI America™

$82.50 - $247.50

Chemical Identifiers

CAS 33904-04-0
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00041075
InChI Key LHPZZVZPOZPDDB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate
PubChem CID 141856
IUPAC Name 4-isothiocyanato-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)N=C=S)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I05415G View Documents TCI America
I05415G
5g Each for $82.50
 
I054125G View Documents TCI America
I054125G
25g Each for $247.50
 
Specifications

Chemical Identifiers

33904-04-0
195.236
LHPZZVZPOZPDDB-UHFFFAOYSA-N
141856
COC1=C(C=C(C=C1)N=C=S)OC
C9H9NO2S
MFCD00041075
3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate
4-isothiocyanato-1,2-dimethoxybenzene

Specifications

White-Yellow
5g
C9H9NO2S
2811
LHPZZVZPOZPDDB-UHFFFAOYSA-N
4-isothiocyanato-1,2-dimethoxybenzene
141856
Crystal-Powder at 20°C
3,4-Dimethoxyphenyl Isothiocyanate
48°C
33904-04-0
MFCD00041075
3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate
COC1=C(C=C(C=C1)N=C=S)OC
195.236
195.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No