missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4-Dimethoxyphenyl Isothiocyanate 98.0+%, TCI America™
Supplier: TCI America I05415G
Specifications
3,4-Dimethoxyphenyl Isothiocyanate | |
48°C | |
C9H9NO2S | |
5 g | |
3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate | |
COC1=C(C=C(C=C1)N=C=S)OC | |
195.236 | |
195.24 | |
Crystalline Powder |
33904-04-0 | |
White-Yellow | |
MFCD00041075 | |
2811 | |
LHPZZVZPOZPDDB-UHFFFAOYSA-N | |
4-isothiocyanato-1,2-dimethoxybenzene | |
141856 | |
≥98.0% (GC) |
Chemical Identifiers
33904-04-0 | |
195.236 | |
LHPZZVZPOZPDDB-UHFFFAOYSA-N | |
141856 | |
COC1=C(C=C(C=C1)N=C=S)OC |
C9H9NO2S | |
MFCD00041075 | |
3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate | |
4-isothiocyanato-1,2-dimethoxybenzene |
Safety and Handling
TSCA : No