Learn More
(3,4-Dimethoxyphenyl)acetic acid, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 115615000
Specifications
(3, 4-Dimethoxyphenyl)acetic acid | |
Plastic bottle | |
White to Beige | |
500g | |
93-40-3 | |
100.0 | |
C10H12O4 | |
MFCD00004335 | |
2-3,4-dimethoxyphenyl acetic acid, 3,4-dimethoxy-phenyl-acetic acid, 3,4-dimethoxybenzeneacetic acid, 3,4-dimethoxyphenyl acetic acid, 3,4-dimethoxyphenylacetic acid, acetic acid, 3,4-dimethoxyphenyl, benzeneacetic acid, 3,4-dimethoxy, homoveratric acid | |
WUAXWQRULBZETB-UHFFFAOYSA-N | |
2-(3,4-dimethoxyphenyl)acetic acid | |
7139 | |
196.2 | |
99% |
Authentic | |
1% max. (K.F.) | |
96°C to 99°C | |
99% | |
98.5 | |
99% | |
(CH3O)2C6H3CH2CO2H | |
10, 409 | |
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents | |
COC1=C(C=C(C=C1)CC(=O)O)OC | |
196.2 | |
CHEBI:86655 | |
Powder |
Chemical Identifiers
93-40-3 | |
196.2 | |
WUAXWQRULBZETB-UHFFFAOYSA-N | |
7139 | |
2-(3,4-dimethoxyphenyl)acetic acid |
C10H12O4 | |
MFCD00004335 | |
2-3,4-dimethoxyphenyl acetic acid, 3,4-dimethoxy-phenyl-acetic acid, 3,4-dimethoxybenzeneacetic acid, 3,4-dimethoxyphenyl acetic acid, 3,4-dimethoxyphenylacetic acid, acetic acid, 3,4-dimethoxyphenyl, benzeneacetic acid, 3,4-dimethoxy, homoveratric acid | |
CHEBI:86655 | |
COC1=C(C=C(C=C1)CC(=O)O)OC |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-244-5
RTECSNumber : AH0675000
TSCA : TSCA