missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3,4-Dimethoxyphenyl)acetic acid, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC115610250

 View more versions of this product

Catalog No. AC115610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(3, 4-Dimethoxyphenyl)acetic acid
Plastic bottle
White to Beige
25g
93-40-3
100.0
C10H12O4
MFCD00004335
2-3,4-dimethoxyphenyl acetic acid
WUAXWQRULBZETB-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)acetic acid
7139
196.2
99%
Authentic
1% max. (K.F.)
96°C to 99°C
99%
98.5
99%
(CH3O)2C6H3CH2CO2H
10, 409
Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
COC1=C(C=C(C=C1)CC(=O)O)OC
196.2
CHEBI:86655
Powder

Chemical Identifiers

93-40-3
196.2
WUAXWQRULBZETB-UHFFFAOYSA-N
7139
2-(3,4-dimethoxyphenyl)acetic acid
C10H12O4
MFCD00004335
2-3,4-dimethoxyphenyl acetic acid
CHEBI:86655
COC1=C(C=C(C=C1)CC(=O)O)OC
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-244-5

RTECSNumber : AH0675000

TSCA : TSCA

SDS Documents