missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxyphenol, 98%

Manufacturer:  Alfa Aesar™ A1134406

 View more versions of this product

Catalog No. AAA1134406



Description

Description

Applications
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
Specifications

Specifications

3,4-Dimethoxyphenol
79°C to 82°C
2033-89-8
MFCD00008390
phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113
COC1=C(C=C(C=C1)O)OC
154.165
154.16
167°C (14mmHg)
5g
C8H10O3
1366650
SMFFZOQLHYIRDA-UHFFFAOYSA-N
3,4-dimethoxyphenol
16251
98%

Chemical Identifiers

2033-89-8
154.165
SMFFZOQLHYIRDA-UHFFFAOYSA-N
16251
COC1=C(C=C(C=C1)O)OC
C8H10O3
MFCD00008390
phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113
3,4-dimethoxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents