missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxyphenethylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC115625000

 View more versions of this product

Catalog No. AC115625000



Description

Description

Specifications

Specifications

3, 4-Dimethoxyphenethylamine
Authentic
1.5450 to 1.5470 (20°C, 589nm)
500g
97.5
98%
(CH3O)2C6H3CH2CH2NH2
13, 800
ANOUKFYBOAKOIR-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)ethanamine
8421
181.23
≥97.5%
1.0700g/mL
Glass bottle
Brown or Orange to Yellow
120-20-7
100.0
C10H15NO2
MFCD00008188
2-3,4-dimethoxyphenyl ethanamine, 2-3,4-dimethoxyphenyl ethylamine, 3,4-dimethoxyphenethylamine, 3,4-dimethoxyphenylethylamine, dimethoxydopamine, dimethoxyphenylethylamine, dimethylmescaline, dimpea, dmpea, homoveratrylamine
COC1=C(C=C(C=C1)CCN)OC
181.235
CHEBI:136995
Viscous Liquid

Chemical Identifiers

120-20-7
181.235
ANOUKFYBOAKOIR-UHFFFAOYSA-N
8421
2-(3,4-dimethoxyphenyl)ethanamine
C10H15NO2
MFCD00008188
2-3,4-dimethoxyphenyl ethanamine, 2-3,4-dimethoxyphenyl ethylamine, 3,4-dimethoxyphenethylamine, 3,4-dimethoxyphenylethylamine, dimethoxydopamine, dimethoxyphenylethylamine, dimethylmescaline, dimpea, dmpea, homoveratrylamine
CHEBI:136995
COC1=C(C=C(C=C1)CCN)OC
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents