Please login to your online account to display your discounted pricing

3,4-Dimethoxyphenethylamine, 98%, ACROS Organics™

$34.52 - $420.80

Chemical Identifiers

Chemical Name or Material 3, 4-Dimethoxyphenethylamine
CAS 120-20-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2
Linear Formula (CH3O)2C6H3CH2CH2NH2
Formula Weight 181.23
MDL Number MFCD00008188
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC115625000

  SDS
thermo fisher scientific
AC115625000
Glass bottle 500g Each for $420.80

AC115620250

  SDS
thermo fisher scientific
AC115620250
Glass bottle 25g Each for $34.52

AC115621000

  SDS
thermo fisher scientific
115621000
Glass bottle 100g Each for $96.28
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 181.23
Color Clear light yellow to orange or slightly brownish
Physical Form Viscous Liquid
Quantity 500g
Density 1.0700g/mL
Beilstein 13, 800
CAS Max % 100.0
Linear Formula (CH3O)2C6H3CH2CH2NH2
Synonym 2-(3, 4-Dimethoxyphenyl)ethylamine
Chemical Name or Material 3, 4-Dimethoxyphenethylamine
CAS 120-20-7
Assay Percent Range 98%
Molecular Formula C10H15NO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00008188
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.