Please login to your online account to display your discounted pricing

3,4-Dimethoxyphenethylamine, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  115621000

 View more versions of this product

Catalog No. AC115621000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Assay ≥97.5%
Formula Weight 181.23
Color Clear light yellow to orange or slightly brownish
Quantity 100g
Beilstein 13, 800
Density 1.0700g/mL
CAS Max % 100.0
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C10H15NO2
Chemical Name or Material 3, 4-Dimethoxyphenethylamine
CAS 120-20-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Infrared Spectrum Authentic
MDL Number MFCD00008188
Synonym 2-(3, 4-Dimethoxyphenyl)ethylamine
CAS Min % 97.5
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2CH2NH2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4-Dimethoxyphenethylamine
CAS 120-20-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2
Linear Formula (CH3O)2C6H3CH2CH2NH2
Formula Weight 181.23
MDL Number MFCD00008188
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.

SDS & Safety Documents