missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxyphenethylamine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC115620250

 View more versions of this product

Catalog No. AC115620250



Specifications

Specifications

3, 4-Dimethoxyphenethylamine
Brown or Orange to Yellow
120-20-7
100.0
Glass bottle
≥97.5%
C10H15NO2
MFCD00008188
Homoveratrylamine, 2-(3, 4-Dimethoxyphenyl)ethylamine
COC1=C(C=C(C=C1)CCN)OC
181.235
CHEBI:136995
Viscous Liquid
1.0700g/mL
25g
97.5
Authentic
1.5450 to 1.5470 (20°C, 589nm)
98%
(CH3O)2C6H3CH2CH2NH2
13, 800
ANOUKFYBOAKOIR-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)ethanamine
8421
181.23

Chemical Identifiers

120-20-7
181.235
ANOUKFYBOAKOIR-UHFFFAOYSA-N
8421
C10H15NO2
MFCD00008188
Homoveratrylamine, 2-(3, 4-Dimethoxyphenyl)ethylamine
CHEBI:136995
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents