missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4'-Dimethoxyflavone, 97%

$70.39 - $223.41

Chemical Identifiers

CAS 4143-62-8
Molecular Formula C17H14O4
Molecular Weight (g/mol) 282.295
MDL Number MFCD00143009
InChI Key ZGHORMOOTZTQFL-UHFFFAOYSA-N
Synonym 2-3,4-dimethoxyphenyl chromen-4-one
PubChem CID 688674
IUPAC Name 2-(3,4-dimethoxyphenyl)chromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL14160MC View Documents Alfa Aesar™
L14160MC
100mg Each for $70.39
 
AAL14160ME View Documents Alfa Aesar™
L14160ME
500mg Each for $223.41
 
Specifications

Chemical Identifiers

4143-62-8
282.295
ZGHORMOOTZTQFL-UHFFFAOYSA-N
688674
COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC
C17H14O4
MFCD00143009
2-3,4-dimethoxyphenyl chromen-4-one
2-(3,4-dimethoxyphenyl)chromen-4-one

Specifications

154°C to 155°C
4143-62-8
MFCD00143009
2-3,4-dimethoxyphenyl chromen-4-one
COC1=C(C=C(C=C1)C2=CC(=O)C3=CC=CC=C3O2)OC
282.295
282.3
3',4'-Dimethoxyflavone
100mg
C17H14O4
236749
ZGHORMOOTZTQFL-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)chromen-4-one
688674
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures