missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 115511000

 View more versions of this product

Catalog No. AC115511000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3, 4-Dimethoxycinnamic acid
Glass bottle
181°C to 184°C
99%
98.5
98.5% min. (GC)
(CH3O)2C6H3CH=CHCO2H
10, 437
HJBWJAPEBGSQPR-GQCTYLIASA-N
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
717531
208.21
99%
Authentic
Yellow to Beige
100g
2316-26-9
100.0
C11H12O4
MFCD00004387
2e-3-3,4-dimethoxyphenyl acrylic acid, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, 3,4-dimethoxycinnamic acid, 3-3,4-dimethoxyphenyl acrylic acid, caffeic acid dimethyl ether, dimethyl caffeic acid, dimethylcaffeic acid, e-3,4-dimethoxycinnamic acid, e-3-3,4-dimethoxyphenyl acrylic acid, unii-bvz841pvjl
COC1=C(C=C(C=C1)C=CC(=O)O)OC
208.21
CHEBI:86549
Powder

Chemical Identifiers

2316-26-9
208.21
HJBWJAPEBGSQPR-GQCTYLIASA-N
717531
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
C11H12O4
MFCD00004387
2e-3-3,4-dimethoxyphenyl acrylic acid, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, 3,4-dimethoxycinnamic acid, 3-3,4-dimethoxyphenyl acrylic acid, caffeic acid dimethyl ether, dimethyl caffeic acid, dimethylcaffeic acid, e-3,4-dimethoxycinnamic acid, e-3-3,4-dimethoxyphenyl acrylic acid, unii-bvz841pvjl
CHEBI:86549
COC1=C(C=C(C=C1)C=CC(=O)O)OC
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-025-5

TSCA : TSCA

SDS Documents