Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxychalcone, 98%

$55.55 - $175.47

Chemical Identifiers

Chemical Name or Material 3,4-Dimethoxychalcone
CAS 53744-28-8
Molecular Formula C17H16O3
Formula Weight 268.32
MDL Number MFCD00041327
Beilstein 2620691
Synonym 2-(3,4-Dimethoxybenzylidene)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1132606

 
alfa aesar™
L1132606
5g Each for $55.55

AAL1132614

 
alfa aesar™
L1132614
25g Each for $175.47
Description & Specifications

Specifications

Formula Weight 268.32
Chemical Name or Material 3,4-Dimethoxychalcone
Synonym 2-(3,4-Dimethoxybenzylidene)acetophenone
Assay 98%
Molecular Formula C17H16O3
Melting Point 88° to 89°C
MDL Number MFCD00041327
CAS 53744-28-8
Beilstein 2620691
Quantity 5g