missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxychalcone, 98%

Manufacturer:  Alfa Aesar™ L1132606

 View more versions of this product

Catalog No. AAL1132606



Specifications

Specifications

3,4-Dimethoxychalcone
5g
C17H16O3
2620691
LSHZPTCZLWATBZ-CSKARUKUSA-N
(E)-3-(3,4-dimethoxyphenyl)-1-phenylprop-2-en-1-one
5354494
98%
88°C to 89°C
53744-28-8
MFCD00041327
3,4-dimethoxychalcone, 3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, e-3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 5, chalcone,4-dimethoxy, e-3,4-dimethoxychalcone, chembl33943, 3,4-dimethoxy benzylidene acetophenone, 2-propen-1-one,4-dimethoxyphenyl-1-phenyl
COC1=C(C=C(C=C1)C=CC(=O)C2=CC=CC=C2)OC
268.312
268.32

Chemical Identifiers

53744-28-8
268.312
LSHZPTCZLWATBZ-CSKARUKUSA-N
5354494
COC1=C(C=C(C=C1)C=CC(=O)C2=CC=CC=C2)OC
C17H16O3
MFCD00041327
3,4-dimethoxychalcone, 3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, e-3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, 2e-3-3,4-dimethoxyphenyl-1-phenylprop-2-en-1-one, chalcone, 5, chalcone,4-dimethoxy, e-3,4-dimethoxychalcone, chembl33943, 3,4-dimethoxy benzylidene acetophenone, 2-propen-1-one,4-dimethoxyphenyl-1-phenyl
(E)-3-(3,4-dimethoxyphenyl)-1-phenylprop-2-en-1-one
Documents