missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Dimethoxybenzylideneacetone, 98+%

$87.59 - $299.18

Chemical Identifiers

CAS 15001-27-1
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00014993
InChI Key XUYBNDHXZMIALN-PLNGDYQASA-N
Synonym 3,4-dimethoxybenzylideneacetone, 4-3,4-dimethoxyphenyl-3-buten-2-one, 3e-4-3,4-dimethoxyphenyl but-3-en-2-one, e-4-3,4-dimethoxyphenyl but-3-en-2-one, 3-buten-2-one, 4-3,4-dimethoxyphenyl, 3,4-dimethoxybenzylideneacetone, 98+%, e-4-3,4-dimethoxyphenyl-3-butene-2-one
PubChem CID 54612014
IUPAC Name (Z)-4-(3,4-dimethoxyphenyl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC(=C(C=C1)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0112106 View Documents Alfa Aesar™
L0112106
5g Each for $87.59
 
AAL0112114 View Documents Alfa Aesar™
L0112114
25g Each for $299.18
 
Specifications

Chemical Identifiers

15001-27-1
206.241
XUYBNDHXZMIALN-PLNGDYQASA-N
54612014
CC(=O)C=CC1=CC(=C(C=C1)OC)OC
C12H14O3
MFCD00014993
3,4-dimethoxybenzylideneacetone, 4-3,4-dimethoxyphenyl-3-buten-2-one, 3e-4-3,4-dimethoxyphenyl but-3-en-2-one, e-4-3,4-dimethoxyphenyl but-3-en-2-one, 3-buten-2-one, 4-3,4-dimethoxyphenyl, 3,4-dimethoxybenzylideneacetone, 98+%, e-4-3,4-dimethoxyphenyl-3-butene-2-one
(Z)-4-(3,4-dimethoxyphenyl)but-3-en-2-one

Specifications

85°C to 87°C
15001-27-1
MFCD00014993
3,4-dimethoxybenzylideneacetone, 4-3,4-dimethoxyphenyl-3-buten-2-one, 3e-4-3,4-dimethoxyphenyl but-3-en-2-one, e-4-3,4-dimethoxyphenyl but-3-en-2-one, 3-buten-2-one, 4-3,4-dimethoxyphenyl, 3,4-dimethoxybenzylideneacetone, 98+%, e-4-3,4-dimethoxyphenyl-3-butene-2-one
CC(=O)C=CC1=CC(=C(C=C1)OC)OC
206.241
206.24
3,4-Dimethoxybenzylideneacetone
5g
C12H14O3
1967327
XUYBNDHXZMIALN-PLNGDYQASA-N
(Z)-4-(3,4-dimethoxyphenyl)but-3-en-2-one
54612014
≥98%