Learn More
3,4-Dimethoxybenzonitrile, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1252122
Description
3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
3,4-Dimethoxybenzonitrile | |
65°C to 69°C | |
MFCD00001802 | |
UN3439 | |
veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole | |
OSEQIDSFSBWXRE-UHFFFAOYSA-N | |
3,4-dimethoxybenzonitrile | |
74842 | |
≥98% |
2024-83-1 | |
C9H9NO2 | |
100 g | |
1451651 | |
Soluble in methanol. | |
COC1=CC=C(C=C1OC)C#N | |
163.18 | |
163.18 |
Chemical Identifiers
2024-83-1 | |
163.18 | |
OSEQIDSFSBWXRE-UHFFFAOYSA-N | |
74842 | |
COC1=CC=C(C=C1OC)C#N |
C9H9NO2 | |
MFCD00001802 | |
veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole | |
3,4-dimethoxybenzonitrile |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 217-969-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only